Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339876.1 | NP_001326797.1 | 4 - 5 | 1284 | 64 | |
NM_115613.4 | NP_191312.2 | 4 - 5 | 1299 | 64 | ||
GenBank mRNA | BT011630.1 | - | 4 - 5 | 1000 | 64 | |
BT012274.1 | - | 4 - 5 | 946 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |