Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036778.2 | NP_001031855.1 | 13 - 14 | 984 | 78 | |
NM_120921.5 | NP_568193.1 | 12 - 13 | 1097 | 78 | ||
GenBank mRNA | AK317201.1 | - | 13 - 14 | 985 | 78 | |
AY085529.1 | - | 12 - 13 | 1013 | 78 | ||
AY090238.1 | - | 12 - 13 | 964 | 78 | ||
BT000619.1 | - | 12 - 13 | 953 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |