Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120958.3 | NP_196484.1 | 2 - 3 | 294 | 78 | |
GenBank mRNA | AY090341.1 | - | 2 - 3 | 259 | 78 | |
X71787.1 | - | 222 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |