Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120958.3 | NP_196484.1 | 6 - 7 | 977 | 143 | |
GenBank mRNA | AY090341.1 | - | 6 - 7 | 942 | 143 | |
X71787.1 | - | 905 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |