Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC64-V | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120979.3 | NP_196504.2 | 5 - 6 | 974 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |