Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121545.5 | NP_197045.1 | 4 - 5 | 1358 | 113 | |
NM_203054.2 | NP_974783.2 | 5 - 6 | 1485 | 113 | ||
GenBank mRNA | AF067798.1 | - | 4 - 5 | 1293 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |