Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2 BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085141.1 | NP_001078610.1 | 3 - 4 | 362 | 123 | |
NM_122100.4 | NP_197592.1 | 3 - 4 | 292 | 123 | ||
NM_203088.2 | NP_974817.1 | 3 - 4 | 255 | 123 | ||
GenBank mRNA | AY128324.1 | - | 3 - 4 | 221 | 123 | |
BT000056.1 | - | 2 - 3 | 150 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |