Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2 BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085141.1 | NP_001078610.1 | 7 - 8 | 720 | 84 | |
NM_122100.4 | NP_197592.1 | 7 - 8 | 650 | 84 | ||
NM_203088.2 | NP_974817.1 | 7 - 8 | 613 | 84 | ||
GenBank mRNA | AY128324.1 | - | 7 - 8 | 579 | 84 | |
BT000056.1 | - | 6 - 7 | 508 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |