Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343870.1 | NP_001318635.1 | 6 - 7 | 838 | 64 | |
NM_001343872.1 | NP_001330710.1 | 6 - 7 | 798 | 64 | ||
NM_122337.5 | NP_197819.2 | 6 - 7 | 874 | 64 | ||
GenBank mRNA | AK222217.1 | - | 6 - 7 | 796 | 64 | |
AK228814.1 | - | 6 - 7 | 676 | 64 | ||
AY817014.1 | - | 5 - 6 | 524 | 64 | ||
BT011734.1 | - | 5 - 6 | 524 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |