Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344404.1 | NP_001318723.1 | 2 - 3 | 1597 | 95 | |
NM_001344405.1 | NP_001332436.1 | 2 - 3 | 1347 | 95 | ||
NM_123525.4 | NP_198976.3 | 3 - 4 | 1615 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |