Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123846.4 | NP_568639.1 | 2 - 3 | 620 | 93 | |
NM_203156.2 | NP_974885.1 | 2 - 3 | 620 | 93 | ||
GenBank mRNA | AK118806.1 | - | 2 - 3 | 576 | 93 | |
AK227530.1 | - | 2 - 3 | 588 | 93 | ||
BT005411.1 | - | 2 - 3 | 501 | 93 | ||
BT006477.1 | - | 2 - 3 | 501 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |