Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344775.1 | NP_001330362.1 | 4 - 5 | 1712 | 58 | |
NM_203178.3 | NP_974907.1 | 5 - 6 | 1202 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |