Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345181.1 | NP_001330467.1 | 6 - 7 | 1023 | 82 | |
NM_125000.4 | NP_568836.2 | 7 - 8 | 902 | 82 | ||
NM_203211.1 | NP_974940.1 | 7 - 8 | 825 | 82 | ||
GenBank mRNA | AF507916.1 | - | 7 - 8 | 795 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |