Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125008.5 | NP_200436.1 | 2 - 3 | 667 | 96 | |
NM_203212.2 | NP_974941.1 | 2 - 3 | 467 | 96 | ||
GenBank mRNA | AY074372.1 | - | 2 - 3 | 499 | 96 | |
AY096754.1 | - | 1 - 2 | 130 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |