Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125204.3 | NP_200627.2 | 5 - 6 | 730 | 139 | |
NM_203225.1 | NP_974954.1 | 5 - 6 | 689 | 139 | ||
GenBank mRNA | AY072018.1 | - | 5 - 6 | 636 | 139 | |
BT000833.1 | - | 5 - 6 | 625 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |