Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345293.1 | NP_001331086.1 | 4 - 5 | 545 | 77 | |
NM_125209.3 | NP_200632.2 | 4 - 5 | 458 | 77 | ||
NM_203228.2 | NP_974957.1 | 3 - 4 | 388 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |