Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125287.5 | NP_568894.1 | 3 - 4 | 649 | 141 | |
NM_203233.1 | NP_974962.1 | 3 - 4 | 615 | 141 | ||
GenBank mRNA | AB021706.1 | - | 3 - 4 | 390 | 141 | |
AY088200.1 | - | 3 - 4 | 546 | 141 | ||
JQ937241.1 | - | 3 - 4 | 375 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |