Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103032.4 | NP_174575.1 | 1 - 2 | 393 | 63 | |
GenBank mRNA | AF325023.2 | - | 1 - 2 | 257 | 63 | |
AF370254.1 | - | 1 - 2 | 257 | 63 | ||
AF412105.1 | - | 1 - 2 | 258 | 63 | ||
AY063070.1 | - | 1 - 2 | 222 | 63 | ||
AY085664.1 | - | 1 - 2 | 261 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |