Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333051.1 | NP_001319134.1 | 4 - 5 | 500 | 83 | |
NM_001333052.1 | NP_001321942.1 | 3 - 4 | 579 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |