Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103079.4 | NP_174620.2 | 4 - 5 | 1863 | 73 | |
GenBank mRNA | AJ581996.1 | - | 4 - 5 | 1539 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |