Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333084.1 | NP_001322412.1 | 8 - 9 | 616 | 85 | |
NM_103129.5 | NP_564436.4 | 8 - 9 | 637 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |