Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333087.1 | NP_001322581.1 | 4 - 5 | 1266 | 76 | |
NM_103135.4 | NP_564437.1 | 8 - 9 | 1555 | 76 | ||
NM_179414.1 | NP_849745.1 | 8 - 9 | 1489 | 76 | ||
NM_202231.1 | NP_973960.1 | 8 - 9 | 1486 | 76 | ||
GenBank mRNA | AK227368.1 | - | 8 - 9 | 1445 | 76 | |
BT006448.1 | - | 8 - 9 | 1424 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |