Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP5PI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333086.1 | NP_001322580.1 | 9 - 10 | 1629 | 65 | |
NM_001333087.1 | NP_001322581.1 | 5 - 6 | 1420 | 65 | ||
NM_103135.4 | NP_564437.1 | 9 - 10 | 1709 | 65 | ||
NM_179414.1 | NP_849745.1 | 9 - 10 | 1643 | 65 | ||
NM_202231.1 | NP_973960.1 | 9 - 10 | 1640 | 65 | ||
GenBank mRNA | AK227368.1 | - | 9 - 10 | 1599 | 65 | |
BT006448.1 | - | 9 - 10 | 1578 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |