Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP94D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103176.2 | NP_174713.1 | 1 | 648 | 127 | |
GenBank mRNA | DQ446321.1 | - | 1 | 648 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |