Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CINV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103255.2 | NP_174791.2 | 5 - 6 | 1508 | 68 | |
NM_179419.4 | NP_849750.1 | 5 - 6 | 1584 | 68 | ||
GenBank mRNA | AM230708.2 | - | 4 - 5 | 1354 | 68 | |
AY065247.1 | - | 5 - 6 | 1507 | 68 | ||
AY142662.1 | - | 4 - 5 | 1354 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |