Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103456.4 | NP_174996.1 | 3 - 4 | 567 | 115 | |
GenBank mRNA | AK230314.1 | - | 3277 | 115 | ||
AY039539.2 | - | 3 - 4 | 439 | 115 | ||
AY039961.1 | - | 3 - 4 | 487 | 115 | ||
AY079402.1 | - | 3 - 4 | 368 | 115 | ||
AY095991.1 | - | 3 - 4 | 433 | 115 | ||
AY140091.1 | - | 3 - 4 | 408 | 115 | ||
BT002267.1 | - | 3 - 4 | 368 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |