Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103466.5 | NP_564483.1 | 1 - 2 | 207 | 66 | |
GenBank mRNA | AK176692.1 | - | 1 - 2 | 186 | 66 | |
AK176760.1 | - | 1 - 2 | 186 | 66 | ||
BT033158.1 | - | 1 - 2 | 111 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons |