Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333208.1 | NP_001321430.1 | 6 - 7 | 821 | 146 | |
NM_001333209.1 | NP_001321431.1 | 5 - 6 | 754 | 146 | ||
NM_103466.5 | NP_564483.1 | 5 - 6 | 641 | 146 | ||
GenBank mRNA | AK176692.1 | - | 5 - 6 | 620 | 146 | |
AK176760.1 | - | 5 - 6 | 620 | 146 | ||
BT033158.1 | - | 5 - 6 | 545 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |