Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103506.1 | NP_175046.1 | 7 - 8 | 1154 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |