Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123962.2 | NP_001117434.1 | 6 - 7 | 1927 | 94 | |
NM_103511.4 | NP_175051.1 | 6 - 7 | 2188 | 94 | ||
GenBank mRNA | AF378782.1 | - | 6 - 7 | 2024 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |