Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084207.1 | NP_001077676.1 | 2 - 3 | 187 | 76 | |
NM_103541.3 | NP_175081.1 | 3 - 4 | 339 | 76 | ||
NM_179439.4 | NP_849770.1 | 2 - 3 | 551 | 76 | ||
GenBank mRNA | AY072122.1 | - | 2 - 3 | 453 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |