Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084207.1 | NP_001077676.1 | 8 - 9 | 983 | 66 | |
NM_103541.3 | NP_175081.1 | 9 - 10 | 1135 | 66 | ||
NM_179439.4 | NP_849770.1 | 8 - 9 | 1347 | 66 | ||
GenBank mRNA | AY072122.1 | - | 8 - 9 | 1249 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |