Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333230.1 | NP_001321683.1 | 4 - 5 | 841 | 80 | |
NM_103548.5 | NP_175088.1 | 5 - 6 | 974 | 80 | ||
NM_202240.2 | NP_973969.1 | 5 - 6 | 927 | 80 | ||
NM_202241.2 | NP_973970.1 | 5 - 6 | 927 | 80 | ||
GenBank mRNA | AK228645.1 | - | 5 - 6 | 892 | 80 | |
AY128357.1 | - | 5 - 6 | 927 | 80 | ||
BT002075.1 | - | 5 - 6 | 855 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |