Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G45110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103586.4 | NP_175126.2 | 8 - 9 | 1014 | 106 | |
GenBank mRNA | AK229448.1 | - | 8 - 9 | 955 | 106 | |
BT010558.1 | - | 8 - 9 | 917 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |