Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103597.5 | NP_175137.1 | 3 - 4 | 488 | 112 | |
NM_179447.2 | NP_849778.1 | 3 - 4 | 532 | 112 | ||
GenBank mRNA | AK221694.1 | - | 3 - 4 | 458 | 112 | |
AY062097.1 | - | 3 - 4 | 434 | 112 | ||
AY090263.1 | - | 3 - 4 | 429 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |