Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA3;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103614.5 | NP_564499.3 | 5 - 6 | 1394 | 89 | |
NM_103615.1 | NP_564500.1 | 5 - 6 | 1134 | 89 | ||
GenBank mRNA | AY064987.1 | - | 5 - 6 | 1129 | 89 | |
AY084924.1 | - | 5 - 6 | 1132 | 89 | ||
AY113013.1 | - | 5 - 6 | 1049 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |