Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KU80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333292.1 | NP_001322094.1 | 2 - 3 | 173 | 108 | |
NM_103701.2 | NP_564520.1 | 2 - 3 | 221 | 108 | ||
GenBank mRNA | AF283758.1 | - | 2 - 3 | 151 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |