Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123974.1 | NP_001117446.1 | 1 - 2 | 777 | 102 | |
NM_103707.5 | NP_175245.1 | 2 - 3 | 376 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |