Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLN1;5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103743.3 | NP_175280.1 | 5 - 6 | 445 | 118 | |
GenBank mRNA | AK118005.1 | - | 5 - 6 | 445 | 118 | |
BT006245.1 | - | 5 - 6 | 372 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |