Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115654.3 | NP_191351.1 | 2 - 3 | 644 | 81 | |
GenBank mRNA | AY086384.1 | - | 2 - 3 | 584 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |