Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339909.1 | NP_001319790.1 | 17 - 18 | 2306 | 81 | |
NM_001339910.1 | NP_001327763.1 | 17 - 18 | 2467 | 81 | ||
NM_001339911.1 | NP_001327764.1 | 17 - 18 | 2467 | 81 | ||
NM_001339912.1 | NP_001327762.1 | 17 - 18 | 2266 | 81 | ||
NM_115678.2 | NP_191375.1 | 16 - 17 | 1955 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |