Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BS14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115679.4 | NP_191376.1 | 4 - 5 | 584 | 67 | |
GenBank mRNA | AF368175.1 | - | 4 - 5 | 370 | 67 | |
AK117351.1 | - | 4 - 5 | 441 | 67 | ||
BT004705.1 | - | 3 - 4 | 275 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |