Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339927.1 | NP_001325607.1 | 4 - 5 | 1879 | 86 | |
NM_115724.4 | NP_191421.2 | 4 - 5 | 1960 | 86 | ||
GenBank mRNA | BT003919.1 | - | 4 - 5 | 1646 | 86 | |
BT005006.1 | - | 4 - 5 | 1590 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |