Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115736.2 | NP_191433.1 | 8 - 9 | 1114 | 122 | |
GenBank mRNA | AY099611.1 | - | 8 - 9 | 1114 | 122 | |
BT002169.1 | - | 8 - 9 | 1050 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |