Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339948.1 | NP_001325700.1 | 1 - 2 | 375 | 71 | |
NM_115765.4 | NP_191462.1 | 1 - 2 | 406 | 71 | ||
GenBank mRNA | AJ294464.1 | - | 1 - 2 | 358 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |