Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339948.1 | NP_001325700.1 | 4 - 5 | 1062 | 62 | |
NM_115765.4 | NP_191462.1 | 4 - 5 | 1093 | 62 | ||
GenBank mRNA | AJ294464.1 | - | 4 - 5 | 1045 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |