Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115767.3 | NP_191464.1 | 5 - 6 | 872 | 88 | |
GenBank mRNA | AY065025.1 | - | 5 - 6 | 854 | 88 | |
AY143905.1 | - | 5 - 6 | 536 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |