Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115767.3 | NP_191464.1 | 7 - 8 | 1073 | 81 | |
GenBank mRNA | AY065025.1 | - | 7 - 8 | 1055 | 81 | |
AY143905.1 | - | 7 - 8 | 737 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |