Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339950.1 | NP_001326589.1 | 7 - 8 | 919 | 98 | |
NM_115772.4 | NP_191469.3 | 13 - 14 | 1947 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |