Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339950.1 | NP_001326589.1 | 32 - 33 | 3898 | 109 | |
NM_115772.4 | NP_191469.3 | 38 - 39 | 4926 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |